Anti-E2F1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3485AEA 360 EUR
ABGEAP3485A
Anti-E2F1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: E2F1 (E2F transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR