Anti-MEF2C Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3385AEA 360 EUR
ABGEAP3385A
Anti-MEF2C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR