Anti-EGFR Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3376AEA 348 EUR
ABGEAP3376A
Anti-EGFR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR