Anti-NR4A1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3339AEA 360 EUR
ABGEAP3339A
Anti-NR4A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NR4A1 (nuclear receptor subfamily 4, group A, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR