Anti-RAF1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3332AEA 348 EUR
ABGEAP3332A
Anti-RAF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAF1 (v-raf-1 murine leukemia viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR