Anti-FOS Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3314AEA 360 EUR
ABGEAP3314A
Anti-FOS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FOS (FBJ murine osteosarcoma viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR