Anti-STAT3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3260AEA 360 EUR
ABGEAP3260A
Anti-STAT3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR