Anti-SEPARIN Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3247AEA 370 EUR
ABGEAP3247A
Anti-SEPARIN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SEPARIN
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR