Anti-RAD9A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3231AEA 348 EUR
ABGEAP3231A
Anti-RAD9A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAD9A (RAD9 homolog A (S. pombe))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR