Anti-ERBB4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3122AEA 370 EUR
ABGEAP3122A
Anti-ERBB4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ERBB4 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR