Anti-CREB Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3077AEA 380 EUR
ABGEAP3077A
Anti-CREB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CREB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR