Anti-CDC25A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3051AEA 348 EUR
ABGEAP3051A
Anti-CDC25A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDC25A (cell division cycle 25A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR