Anti-BIK Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3042AEA 348 EUR
ABGEAP3042A
Anti-BIK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BIK (BCL2-interacting killer (apoptosis-inducing))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR