Anti-BAD Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3035AEA 360 EUR
ABGEAP3035A
Anti-BAD Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BAD (BCL2-associated agonist of cell death)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR