Anti-HIST3H3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3004AEA 360 EUR
ABGEAP3004A
Anti-HIST3H3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR