Anti-MDH2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2909CEA 370 EUR
ABGEAP2909C
Anti-MDH2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MDH2 (malate dehydrogenase 2, NAD (mitochondrial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR