Anti-DDAH1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2898BEA 360 EUR
ABGEAP2898B
Anti-DDAH1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDAH1 (dimethylarginine dimethylaminohydrolase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR