Anti-AHSA1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2892AEA 348 EUR
ABGEAP2892A
Anti-AHSA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AHSA1 (AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR