Anti-EXO1 Rabbit Polyclonal Antibody
Lieferant: Abgent
ABGEAP2871AEA 350 EUR
ABGEAP2871A
Anti-EXO1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EXO1 (exonuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human