Anti-LSM1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2863CEA 348 EUR
ABGEAP2863C
Anti-LSM1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LSM1 (LSM1, U6 small nuclear RNA associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR