Anti-INPP5D Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2845CEA 350 EUR
ABGEAP2845C
Anti-INPP5D Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: INPP5D (inositol polyphosphate-5-phosphatase, 145kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR