Anti-U2AF1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2802CEA 350 EUR
ABGEAP2802C
Anti-U2AF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: U2AF1 (U2 small nuclear RNA auxiliary factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR