Anti-RFC2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2797AEA 350 EUR
ABGEAP2797A
Anti-RFC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RFC2 (replication factor C (activator 1) 2, 40kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR