Anti-M6PR Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2792BEA 348 EUR
ABGEAP2792B
Anti-M6PR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: M6PR (mannose-6-phosphate receptor (cation dependent))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR