Anti-NSE Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2780BEA 350 EUR
ABGEAP2780B
Anti-NSE Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NSE
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR