Anti-HNF4A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2778AEA 348 EUR
ABGEAP2778A
Anti-HNF4A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HNF4A (hepatocyte nuclear factor 4, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR