Anti-ITGA6 (isoform 2) Rabbit Polyclonal Antibody
Lieferant: Abgent
ABGEAP2753AEA 350 EUR
ABGEAP2753A
Anti-ITGA6 (isoform 2) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ITGA6 (isoform 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human