Anti-KLF4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2725FEA 360 EUR
ABGEAP2725F
Anti-KLF4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLF4 (Kruppel-like factor 4 (gut))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR