Anti-CCND1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2612CEA 360 EUR
ABGEAP2612C
Anti-CCND1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCND1 (cyclin D1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR