Anti-MECP2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2545AEA 350 EUR
ABGEAP2545A
Anti-MECP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MECP2 (methyl CpG binding protein 2 (Rett syndrome))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR