Anti-NUB1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2510AEA 350 EUR
ABGEAP2510A
Anti-NUB1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NUB1 (negative regulator of ubiquitin-like proteins 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR