Anti-PIGK Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2462BEA 350 EUR
ABGEAP2462B
Anti-PIGK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIGK (phosphatidylinositol glycan anchor biosynthesis, class K)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR