Anti-AGL Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2402BEA 360 EUR
ABGEAP2402B
Anti-AGL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AGL (amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR