Anti-SYVN1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2184EEA 360 EUR
ABGEAP2184E
Anti-SYVN1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SYVN1 (synovial apoptosis inhibitor 1, synoviolin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR