Anti-Mib1/Mindbomb Rabbit Polyclonal Antibody
Lieferant: Abgent
ABGEAP2172AEA 348 EUR
ABGEAP2172A
Anti-Mib1/Mindbomb Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Mib1/Mindbomb
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human