Anti-UCHL3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2127BEA 370 EUR
ABGEAP2127B
Anti-UCHL3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UCHL3 (ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR