Anti-UCHL1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2126AEA 350 EUR
ABGEAP2126A
Anti-UCHL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR