Anti-UBE2L3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2117BEA 348 EUR
ABGEAP2117B
Anti-UBE2L3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UBE2L3 (ubiquitin-conjugating enzyme E2L 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR