Anti-SMURF2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2105AEA 360 EUR
ABGEAP2105A
Anti-SMURF2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SMURF2 (SMAD specific E3 ubiquitin protein ligase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR