Anti-PROX-1-S514 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2035EEA 360 EUR
ABGEAP2035E
Anti-PROX-1-S514 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PROX-1-S514
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR