Anti-NEUROG2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2023BEA 350 EUR
ABGEAP2023B
Anti-NEUROG2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NEUROG2 (neurogenin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR