Anti-BMPR1A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP2004FEA 360 EUR
ABGEAP2004F
Anti-BMPR1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BMPR1A (bone morphogenetic protein receptor, type IA)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR