Anti-EIF4EBP1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1981CEA 350 EUR
ABGEAP1981C
Anti-EIF4EBP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR