Anti-LEO1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1978AEA 350 EUR
ABGEAP1978A
Anti-LEO1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LEO1 (Leo1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR