Anti-TCF7 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1951AEA 360 EUR
ABGEAP1951A
Anti-TCF7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TCF7 (transcription factor 7 (T-cell specific, HMG-box))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR