Anti-RPL23A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1939CEA 348 EUR
ABGEAP1939C
Anti-RPL23A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPL23A (ribosomal protein L23a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR