Anti-CSE1L Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1935AEA 370 EUR
ABGEAP1935A
Anti-CSE1L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CSE1L (CSE1 chromosome segregation 1-like (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR