Anti-UPF1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1905BEA 348 EUR
ABGEAP1905B
Anti-UPF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UPF1 (UPF1 regulator of nonsense transcripts homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR