Anti-GABARAPL2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1822DEA 360 EUR
ABGEAP1822D
Anti-GABARAPL2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GABARAPL2 (GABA(A) receptor-associated protein-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR