Anti-RGS19 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1820FEA 350 EUR
ABGEAP1820F
Anti-RGS19 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RGS19 (regulator of G-protein signaling 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR